Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Faecal containers with screw cap

25 ml, with spoon, without label, PP, 25 mm, 80 mm
Catalog # LPIT109048
Supplier:   LP ITALIANA
Faecal containers with screw cap
Catalog # LPIT109048
Supplier:  LP ITALIANA
Faecal containers with screw cap
Faecal containers with screw cap

25 ml, with spoon, without label, PP, 25 mm, 80 mm

Price
1.770,00 kr
Log in for availability
1 * 1.000 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

Containers made of PP, translucent.

  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean

With or without spoon and label.

 : *The product(s) on this page are outside the scope of the EU Directive 2019/904

 

Specifications
  • Ext.Ø:
    25 mm
  • Type:
    with spoon, without label
  • Capacity:
    25 ml
  • Height:
    80 mm
  • Material:
    PP