Polystyrene standards, GPC/SEC columns calibration
Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g, 1
Catalog # VARIPL2013-9005
Supplier: VARIAN
Polystyrene standards, GPC/SEC columns calibration
Catalog # VARIPL2013-9005
Supplier: VARIAN
CAS Number:
Jump to:
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
- Compatible with most organic solvents
- Certificate of analysis meets international protocols
- Calibration capability for virtually all applications
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:1
- Description:Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g