Order Entry
Denmark
Orders LinkContactUsLinkComponent
Polystyrene standards, GPC/SEC columns calibration
Polystyrene standards, GPC/SEC columns calibration
Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g, 1
Catalog # VARIPL2013-8005
Supplier:   VARIAN
Polystyrene standards, GPC/SEC columns calibration
Catalog # VARIPL2013-8005
Supplier:  VARIAN
CAS Number:  
Polystyrene standards, GPC/SEC columns calibration
Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g, 1
CAS Number:  
Price
7.080,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
  • Compatible with most organic solvents
  • Certificate of analysis meets international protocols
  • Calibration capability for virtually all applications
Documents
Specifications
Specifications
  • Pk:
    1
  • Description:
    Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g
Specifications
Anbefalinger