Order Entry
Denmark
Orders LinkContactUsLinkComponent
Molecular models: RNA, Molymod®
Molecular models: RNA, Molymod®
Molecular model of RNA
Catalog # SPIRRNA-24
Marketsource
Supplier:   SPIRING ENTERPRISES
Molecular models: RNA, Molymod®
Catalog # SPIRRNA-24
Marketsource
Molecular models: RNA, Molymod®
Molecular models: RNA, Molymod®
Molecular model of RNA
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
The Molymod system is the original dual-scale system of high quality molecular models.
Delivery information: Protein synthesis kit with: 24 bases, 12 ribose/phophate, 4 TRNA, 4 amino acid parts.
Specifications
  • Description:
    Molecular model of RNA
  • Seq:
    001