Order Entry
Denmark
ContactUsLinkComponent
Panasonic CO2 Incubator with Safe Cell UV fitted, 170 litres. ? InCuSafe copper enriched Stainless Steel Interior for continuous surface decontamination. Automatically decontaminates all-day-e very-day without the need to run dedicated decontaminat 1 * 1 items
Catalog # SANYMCO-19AICUV
Supplier:  PHC Corporation
Panasonic CO2 Incubator with Safe Cell UV fitted, 170 litres. ? InCuSafe copper enriched Stainless Steel Interior for continuous surface decontamination. Automatically decontaminates all-day-e very-day without the need to run dedicated decontaminat 1 * 1 items
Catalog # SANYMCO-19AICUV
Supplier:  PHC Corporation
Frequently Bought Together