Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spore strips
Spore strips
Geobacillus stearothermophilus cell line 7953: traditional spore strip 107 (24 month shelf life) / sterilisation process: steam, 30, 6 mm
Catalog # NAMSBS-108
Supplier:   CROSSTEX
Spore strips
Catalog # NAMSBS-108
Supplier:  CROSSTEX
Spore strips
Geobacillus stearothermophilus cell line 7953: traditional spore strip 107 (24 month shelf life) / sterilisation process: steam, 30, 6 mm
Price
Inquire For Price
Log in for availability
1 * 100 Items
Product Details & Documents
Documents
Specifications
  • Width:
    6 mm
  • Description:
    Geobacillus stearothermophilus cell line 7953: traditional spore strip 107 (24 month shelf life) / sterilisation process: steam
  • Length:
    30
  • Seq:
    03