Order Entry
Denmark
ContactUsLinkComponent
Lc/ms nitrogen generators (psa) for nebulisation, sheath, drying, curtain, exhaust and collision cell applications, 230v, n2: 20l/mi n, >99%*. air: 3l/min, (-40c dp). 7 bar g (101.5 psi g), without compressor 1 * 1 items
Catalog # DHUNLCMS12-2-0-E
Supplier:  DOMNICK HUNTER
Lc/ms nitrogen generators (psa) for nebulisation, sheath, drying, curtain, exhaust and collision cell applications, 230v, n2: 20l/mi n, >99%*. air: 3l/min, (-40c dp). 7 bar g (101.5 psi g), without compressor 1 * 1 items
Catalog # DHUNLCMS12-2-0-E
Supplier:  DOMNICK HUNTER
Frequently Bought Together