Order Entry
Denmark
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, HyperSep™ SCX

Solid phase extraction cartridges, HyperSep™ SCX

Cartridge, SCX, 40 - 60 µm, 1 ml, 100 mg
Catalog # HYPE60108-421
Solid phase extraction cartridges, HyperSep™ SCX
Catalog # HYPE60108-421
Solid phase extraction cartridges, HyperSep™ SCX
Solid phase extraction cartridges, HyperSep™ SCX

Cartridge, SCX, 40 - 60 µm, 1 ml, 100 mg

Price
2.680,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents

A strong cation exchange sorbent for extraction of charged basic compounds.

  • Benzene ring provides potential for nonpolar interactions
  • Low pKa value sorbent
Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    1 ml
  • Seq:
    02
  • Bed weight:
    100 mg
  • Phase:
    SCX