Order Entry
Denmark
Orders LinkContactUsLinkComponent
Extender tools
Extender tools
Extender tool, Aluminium, Headless knurl
Catalog # UPCHP-297
Supplier:   UPCHURCH SCIENTIFIC
Extender tools
Catalog # UPCHP-297
Extender tools
Extender tools
Extender tool, Aluminium, Headless knurl
Price
211,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.
Specifications
  • Description:
    Extender tool
  • Material:
    Aluminium
  • Seq:
    05
  • Use with head style:
    Headless knurl