Order Entry
Denmark
Orders LinkContactUsLinkComponent
Draining racks, wall mounted
Draining rack, with 16 rods of 200 mm and 16 rods of 140 mm, 450×600 mm
  SEMA2110
 :   SEMADENI
Draining racks, wall mounted
  SEMA2110
 :  SEMADENI
Draining racks, wall mounted
Draining rack, with 16 rods of 200 mm and 16 rods of 140 mm, 450×600 mm
3.450,00 kr
1 * 1 Items
White, PVC drain channel, polyethylene coated metal rack.
  • Drain channels inclined toward the centre and provided with a drain point
  • Metal frame and drip plasticised PE rods
  • The rack is hung on the wall plate and can be easily removed for cleaning
  • Description:
    Draining rack, with 16 rods of 200 mm and 16 rods of 140 mm
  • Seq:
    02
  • W×H:
    450×600 mm