Trays, multipurpose, PP
White, PP, 535×635×140 mm
Catalog # SEMA3618
Supplier: SEMADENI
Jump to:
Product Details & Documents
Made of PP, white.
- Stable, durable, unbreakable
- Temperature resistant from −20 to +130 °C
- Smooth surfaces for easy cleaning
Multipurpose trays. Suitable for use as a tray for aggressive substances, sterilisation and photographic development.
Documents
SEMA3618_TDS_EN
Datasheet
Specifications
- Colour:White
- Material:PP
- W×D×H:535×635×140 mm



