Order Entry
Denmark
Orders LinkContactUsLinkComponent
Corning® DeckWorks Reload Systems
Corning® DeckWorks Reload Systems
Low binding, 1 - 300 µl, -, Transparent, 960 tips/pack
Catalog # 613-1519
   Waste Reducing
Supplier:   Corning
Corning® DeckWorks Reload Systems
Catalog # 613-1519
Supplier:  Corning
   Waste Reducing
Corning® DeckWorks Reload Systems
Corning® DeckWorks Reload Systems
Low binding, 1 - 300 µl, -, Transparent, 960 tips/pack
Price
2.630,00 kr
Log in for availability
1 * 3.840 Items
Product Details & Documents
Corning® DeckWorks™ pipette tip system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials.
  • Certified free of RNase, DNase, pyrogens and PCR inhibitors
Corning DeckWorks reload systems are available in 10, 200, 300 and 1000 µl volumes and allow for convenient and economical reloading of durable Corning DeckWorks hinged racks. DeckWorks reload decks are packaged in transparent trays for easy tip identification, require no transfer devices, save space and minimize packaging waste. Although the pipette tips are made from virgin medical-grade resins, the reload decks and rack bases are manufactured from 100% recycled polypropylene. Non sterile decks and racks are steam autoclavable.
Corning DeckWorks low binding tips are available in 10, 200 and 300 µl sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
Both the hinged racks and relaod decks within this system are manufactured with recycled plastic. The entire DeckWorks system is comprised of 7 sizes of graduated pipette tips which are longer than standard tips to minimize the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300 and 1000 µl tips. The pipette tip s are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. Medical grade resins and production excellence allow for full certification as: nonpyrogenic, human DNA-free, RNase-/DNase-free, sterility assuarance level 10⁻³. All tips are manufactured from polypropylene.
Ordering information: Corning DeckWorks pipette tips are also available in bulk, hinged rack, barrier hinged rack, reload starter kit, and tip station configurations.
Specifications
  • Packed:
    960 tips/pack
  • Sterile:
    -
  • Volume:
    1 - 300 µl
  • Tip style:
    Low binding
  • Tip colour:
    Transparent
  • Cat. No.:
    613-1519
  • Environmentally Preferable:
    Waste Reducing