Formamide solution, NMR reference standard, Supelco®
Formamide solution, 90% in DMSO-d6 (99.9 atom % D), NMR reference standard, NMR tube size 5 mm x 8 in., 1
Katalog # 551384-8IN
Leverandør: Merck
Formamide solution, NMR reference standard, Supelco®
Katalog # 551384-8IN
Leverandør: Merck
CAS nummer:
Jump to:
Product Details & Documents
Specifications
Specifications
- Concentration:90% in DMSO-d6 (99.9 atom % D)
- Pk:1
- Standards:NMR reference standard
- Description:Formamide solution
- Pk Info:NMR tube size 5 mm x 8 in.
- Cat. No.:551384-8IN
- Description:Formamide solution



