Order Entry
Denmark
Orders LinkContactUsLinkComponent
Plasmid Midiprep kit, GeneJET™

Plasmid Midiprep kit, GeneJET™

GeneJET™ Plasmid Midiprep Kit, 100 preps, 1 KIT
Catalog # K0482
Plasmid Midiprep kit, GeneJET™
Catalog # K0482
CAS Number:  
Plasmid Midiprep kit, GeneJET™
Plasmid Midiprep kit, GeneJET™

GeneJET™ Plasmid Midiprep Kit, 100 preps, 1 KIT

CAS Number:  
Price
6.530,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

GeneJET™ Plasmid Midiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures. The kit utilises silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 200 µg of high copy number plasmid DNA. The purified plasmid DNA can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

  • Efficient: High yields up to 200 µg of high quality plasmid DNA from 50 ml of bacterial culture
  • High concentration of DNA purified: >400 ng/µl
  • Convenient: Spin column format, centrifuge or vacuum protocols
  • Fast: Procedure takes 60 minutes, considerably faster than using gravity flow columns

Delivery information: Each kit includes resuspension solution, lysis solution, neutralisation solution, endotoxin binding reagent, Wash Solution I (concentrated), Wash Solution II (concentrated), RNase A solution, elution buffer (10 mM Tris-HCl, pH 8,5), GeneJET™ Midi purification columns pre-assembled with collection tubes (15 ml), and collection tubes (15 ml).

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    GeneJET™ Plasmid Midiprep Kit, 100 preps
  • Cat. No.:
    K0482
  • Prep size:
    Midi
  • Refine target molecule:
    Plasmid
  • Target molecule:
    Plasmid
  • Method / Format:
    Column (silica-based)
  • Description:
    GeneJET™ Plasmid Midiprep Kit
  • Sample type:
    Bacteria

Specifications