Order Entry
Denmark
Orders LinkContactUsLinkComponent
Screw caps for micro tubes
Screw caps for micro tubes
Screw caps for micro tubes, Natural
Catalog # SARS65.716
Marketsource
Supplier:   SARSTEDT
Screw caps for micro tubes
Catalog # SARS65.716
Supplier:  SARSTEDT
Marketsource
Screw caps for micro tubes
Screw caps for micro tubes, Natural
Price
Inquire For Price
Log in for availability
1 * 1.000 Items
Product Details & Documents
PP, coloured.
  • Excellent chemical resistance
  • Excellent temperature resistance
Screw cap with O-ring seal.
Specifications
  • Colour:
    Natural
  • Description:
    Screw caps for micro tubes
  • Seq:
    06