TE Buffers 100X
TE buffer, 100X, 8,0, Plastic bottle, 250 mL
Catalog # 786-752
Supplier: G-Biosciences
Jump to:
Product Details & Documents
TE Buffer (Tris-EDTA Buffer) is a common molecular biology buffer that is used to protect DNA and RNA from degradation. The Tris buffering agent and EDTA metal chelating properties help protect DNA and RNA.
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:250 mL
- Description:TE buffer, 100X
- pH value:8,0