Order Entry
Denmark
ContactUsLinkComponent
Plate 96-well Well-Coated Protein G clear designed to bind the constant (Fc) region of immunoglobulins ensuring that the antigen bin ding domain of the antibody is orientated away from the plate, offering maximum exposure of the binding site.  The im 1 * 5 items
  786-732
Plate 96-well Well-Coated Protein G clear designed to bind the constant (Fc) region of immunoglobulins ensuring that the antigen bin ding domain of the antibody is orientated away from the plate, offering maximum exposure of the binding site.  The im 1 * 5 items
  786-732
Frequently Bought Together