Order Entry
Denmark
Orders LinkContactUsLinkComponent
Blue Dextran loading dye

Blue Dextran loading dye

3×1,5 ml, 3
Catalog # 786-557
Supplier:   G-Biosciences
Blue Dextran loading dye
Catalog # 786-557
Supplier:  G-Biosciences
CAS Number:  87915-38-6
Blue Dextran loading dye
Blue Dextran loading dye

3×1,5 ml, 3

CAS Number:  87915-38-6
Price
653,00 kr
Log in for availability
1 * 3 Items
Product Details & Documents

A proprietary mixture of Formaldehyde, EDTA and Blue Dextran that is designed for use in loading DNA samples onto denaturing polyacrylamide gels.

Commonly used in loading DNA samples onto denaturing polyacrylamide gels.

Synonyms:  Dextran, blue
  • CAS Number:87915-38-6
  • Formula:(C₂₉H₂₁N₇O₁₂S₃·3Na)n
  • MDL Number:MFCD00130661
Specifications

Specifications

  • Pk:
    3
  • Pk Info:
    3×1,5 ml

Specifications