Vent,barrel,for female thread : R 3/4,membrane diameter : 29 mm,thickness of membrane : 0.2 mm,thread of cap : GL 32. 1 * 1 items
BOHLN1696-32
Vent,barrel,for female thread : R 3/4,membrane diameter : 29 mm,thickness of membrane : 0.2 mm,thread of cap : GL 32. 1 * 1 items
BOHLN1696-32
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection