Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Canister with stand for holding any trap insert, clear polypropylene canister allows visual inspection of trap contents
Catalog # LBCP7815301
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP7815301
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Canister with stand for holding any trap insert, clear polypropylene canister allows visual inspection of trap contents
Price
4.510,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Canister with stand for holding any trap insert, clear polypropylene canister allows visual inspection of trap contents
  • Seq:
    23