Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Oxygen and moisture removal tubing kit, incl. 25' fluorinated ethylene propylene (FEP) tubing, eight tube inserts and two connectors to connect the Oxygen and Moisture Removal Column to diaphragm pump
Catalog # LBCP5242501
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5242501
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Oxygen and moisture removal tubing kit, incl. 25' fluorinated ethylene propylene (FEP) tubing, eight tube inserts and two connectors to connect the Oxygen and Moisture Removal Column to diaphragm pump
Price
5.470,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Oxygen and moisture removal tubing kit, incl. 25' fluorinated ethylene propylene (FEP) tubing, eight tube inserts and two connectors to connect the Oxygen and Moisture Removal Column to diaphragm pump
  • Seq:
    22