Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Tissue holder, epoxy-coated steel, accommodates a box of tissues in the upside down or upright position; opening in the holder permits wipes to be dispensed from the bottom
Catalog # LBCP3927800
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP3927800
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Tissue holder, epoxy-coated steel, accommodates a box of tissues in the upside down or upright position; opening in the holder permits wipes to be dispensed from the bottom
Price
996,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Tissue holder, epoxy-coated steel, accommodates a box of tissues in the upside down or upright position; opening in the holder permits wipes to be dispensed from the bottom
  • Seq:
    42