Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Bottle holder, epoxy-coated steel, with 3" diameter opening, holds a squirt bottle or other round container
Catalog # LBCP3927700
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP3927700
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Bottle holder, epoxy-coated steel, with 3" diameter opening, holds a squirt bottle or other round container
Price
694,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Bottle holder, epoxy-coated steel, with 3" diameter opening, holds a squirt bottle or other round container
  • Seq:
    41