Accessories for glove boxes, Precise™
Electrical pass-through kit, provides a leak-tight connection for data transfer to a printer or computer; incl. sealed 9 pin connector (D-subminiature with male pins on one side and female pins on the other) and 10' cord; requires installation and drill hole; Installation Kit 5241100 is recommended
Catalog # LBCP5075619
Supplier: LABCONCO
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Electrical pass-through kit, provides a leak-tight connection for data transfer to a printer or computer; incl. sealed 9 pin connector (D-subminiature with male pins on one side and female pins on the other) and 10' cord; requires installation and drill hole; Installation Kit 5241100 is recommended
- Seq:32