Order Entry
Denmark
Orders LinkContactUsLinkComponent
Boiling baths, SBB Aqua Plus series
Boiling baths, SBB Aqua Plus series
SBB Aqua 12 Plus, 100 °C, 1,35/1,5 kW, 12 L, 290×315×125 mm, 390×385×440 mm
Catalog # 462-0166
Supplier:   GRANT INSTRUMENTS
Boiling baths, SBB Aqua Plus series
Catalog # 462-0166
Supplier:  GRANT INSTRUMENTS
Boiling baths, SBB Aqua Plus series
Boiling baths, SBB Aqua Plus series
SBB Aqua 12 Plus, 100 °C, 1,35/1,5 kW, 12 L, 290×315×125 mm, 390×385×440 mm
Price
12.350,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Boiling baths are designed for continuous operation with water above 90 °C. They have a stainless steel tank in an outer case and analogue control by means of a simple dial for straightforward temperature settings. Baths have two fixed over-temperature safety cut-outs which can be reset by the user.
  • Energy regulator can be adjusted to ensure a steady boiling action, avoiding excessive steam or spitting
  • Constant level device maintains the required liquid level, avoiding the need to monitor and top up the bath
  • Heater and temperature sensors are mounted at the base of the tank, protected by a perforated tray, giving more usable space
  • Robust construction, durable and resistant to corrosion
  • New modern design
Required accessories must be ordered separately, see SUB Aqua Plus baths entry.
Ordering information: Required accessories must be ordered separately, see SUB Aqua Pro baths entry.
Delivery information: Supplied with polycarbonate lid but without rings.
Caution: ** Required if flat-bottomed flasks are to be placed directly onto the base of the bath and to allow better convection in the bath.
Specifications
  • Heating capacity:
    1,35/1,5 kW
  • Temp.:
    100 °C
  • Capacity:
    12 L
  • Seq:
    02
  • W×D×H:
    390×385×440 mm
  • Bath W×D×H:
    290×315×125 mm
  • Type:
    SBB Aqua 12 Plus
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel rack for 1.5 ml microtubes
    Stainless steel rack for 1.5 ml microtubes
    462-8458
    65 tubes
    Log in for availability
    1 * 1 Items - 1.250,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel rack for 0.5 ml microtubes
    Stainless steel rack for 0.5 ml microtubes
    462-8457
    105 tubes
    Log in for availability
    1 * 1 Items - 1.270,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel base tray**
    Stainless steel base tray**
    462-8350
    SAP12, JBN12, JBA12, GLS Aqua 12 Plus , SBB Aqua 12 Plus
    Log in for availability
    1 * 1 Items - 961,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel base tray**
    Stainless steel base tray**
    462-8351
    SAP18/26. JBN18/26, JBA18, GLS Aqua 18 Plus, OLS200 , SBB Aqua 18/26 Plus
    Log in for availability
    1 * 1 Items - 1.030,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 30 mm Ø tubes
    Stainless steel test tube rack for 30 mm Ø tubes
    462-8456
    12 tubes
    Log in for availability
    1 * 1 Items - 1.250,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 19 mm Ø tubes
    Stainless steel test tube rack for 19 mm Ø tubes
    462-8454
    32 tubes
    Log in for availability
    1 * 1 Items - 1.250,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 25 mm Ø tubes
    Stainless steel test tube rack for 25 mm Ø tubes
    462-8455
    18 tubes
    Log in for availability
    1 * 1 Items - 1.250,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 16 mm Ø tubes
    Stainless steel test tube rack for 16 mm Ø tubes
    462-8453
    36 tubes
    Log in for availability
    1 * 1 Items - 1.250,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 13 mm Ø tubes
    Stainless steel test tube rack for 13 mm Ø tubes
    462-8452
    55 tubes
    Log in for availability
    1 * 1 Items - 1.250,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 10 mm Ø tubes
    Stainless steel test tube rack for 10 mm Ø tubes
    462-8451
    84 tubes
    Log in for availability
    1 * 1 Items - 1.250,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Gabled stainless steel lid
    Gabled stainless steel lid
    462-8212
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26, GLS Aqua 18 Plus), LSB18
    Log in for availability
    1 * 1 Items - 2.460,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Gabled stainless steel lid
    Gabled stainless steel lid
    462-8211
    JBA12, JBN12, SAP12, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus, GLS Aqua 12 Plus), LSB12
    Log in for availability
    1 * 1 Items - 2.190,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Gabled stainless steel lid
    Gabled stainless steel lid
    462-8210
    JBA5, JBN5, SAP2S, SAP5, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    Log in for availability
    1 * 1 Items - 2.000,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Flat lid with 6 ring sets
    Flat lid with 6 ring sets
    462-8222
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26 Plus)
    Log in for availability
    1 * 1 Items - 5.080,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Flat lid with 4 ring sets
    Flat lid with 4 ring sets
    462-8221
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    Log in for availability
    1 * 1 Items - 3.670,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Raised half shelf with perforated mesh
    Raised half shelf with perforated mesh
    462-8232
    JBN26, SAP26, (SUB Aqua 26, JB Aqua 26, SBB Aqua 26 Plus)
    Log in for availability
    1 * 1 Items - 2.130,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Flat lid with 2 ring sets
    Flat lid with 2 ring sets
    462-8220
    JBA5, JBN5, SAP5, SAPD, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    Log in for availability
    1 * 1 Items - 2.380,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Raised half shelf with perforated mesh
    Raised half shelf with perforated mesh
    462-0187
    JBA18, JBN18, SAP18, (SUB Aqua 18, JB Aqua 18, SBB Aqua 18 Plus)
    Log in for availability
    1 * 1 Items - 1.790,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Raised half shelf with perforated mesh
    Raised half shelf with perforated mesh
    462-8231
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    Log in for availability
    1 * 1 Items - 1.520,00 kr
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement lids, polycarbonate
    Replacement lids, polycarbonate
    GRANAQL5
    SAP2S, SAP5 , JBN5, JBA5, SBB Aqua 5 Plus
    Log in for availability
    1 * 1 Items - 1.020,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Polycarbonate lid
    Polycarbonate lid
    462-0333
    Log in for availability
    1 * 1 Items - 1.250,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Polycarbonate lid
    Polycarbonate lid
    462-0334
    Log in for availability
    1 * 1 Items - 1.420,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Stainless steel base tray**, for SAP2S, SAP5, JBN5, JBA5 and SBB Aqua 5 Plus
    Stainless steel base tray**, for SAP2S, SAP5, JBN5, JBA5 and SBB Aqua 5 Plus
    462-8131
    Log in for availability
    1 * 1 Items - 1.030,00 kr
Product Information
  • Catalog #
    Material
    Ø
    Availability
    Price
  • Anti-evaporation spheres
    Anti-evaporation spheres
    462-8239
    PP
    20 mm
    Log in for availability
    1 * 300 Items - 1.030,00 kr