GAS INLET FILTER 0.3µM 1 * 1 items
Catalog # HERA770001
Supplier: Thermo Scientific
Frequently Bought Together
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Lithium Polymer Rechargable Battery
Supplier: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Magnetic stirring bars, cylindrical
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Cover gasket 1 * 1 items
Supplier: VWR Collection
Disposable weighing dishes
Supplier: LUDI
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
FILTRE HEPA FORMA 3311
Supplier: THERMO HYBAID
REPLACEMENT MAIN HEPA FILTER 1 * 1 items
Supplier: Thermo Scientific
Accessories for CBM910 impact printers B+S
Supplier: BELLINGHAM STANLEY