Order Entry
Denmark
Orders LinkContactUsLinkComponent
Selfpacker® Preparative HPLC Columns
O-ring set for frit holder NW100, — mm, — mm
Catalog # 556-0028
Selfpacker® Preparative HPLC Columns
Catalog # 556-0028
Selfpacker® Preparative HPLC Columns
Selfpacker® Preparative HPLC Columns
O-ring set for frit holder NW100, — mm, — mm
Price
2.670,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
This column-packing technology is based on a combination of vacuum suction with subsequent column bed compression that stabilises the packing. A common water-jet pump is used to generate the vacuum. The sorbent slurry is sucked through a packing reservoir into the column by applying low pressure at the bottom end of the column.
  • Allows precise adjustment of the desired packing volume or sorbent bed height
  • Facilitates the determination of the column geometry
  • Makes direct scale-up from analytical to preparative scale easy
Packing stand NW25
Min./max. packing height: 185/220 mm
Max. working pressure: 200 bar
Packing stand NW50
Min./max. packing height: 185/220 mm
Max. working pressure: 100 bar
Packing stand NW100
Min./max. packing height: 295/350 mm
Max. working pressure: 100 bar
Max. working pressure: 200 bar
Specifications
  • Ø int.:
    — mm
  • Description:
    O-ring set for frit holder NW100
  • Length:
    — mm
  • Seq:
    56