Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Pale orange
Catalog # 720-1938
Supplier: Simport Scientific
Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Catalog # 720-1938
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Acetal polymer. Sleeved cassettes especially made to be used with Thermo Fisher printers. Cassettes stacked with tape should be used with Leica and Sakura inkjet printers.
- The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage
- Keep specimens safely submerged
- Totally resistant to the chemical action of solvents used in histology laboratories
Specifications
- Colour:Pale orange



