Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T

Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T

Purple
Catalog # 720-1922
Supplier:   Simport Scientific
Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Catalog # 720-1922
Supplier:  Simport Scientific
Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T

Purple

Price
1.770,00 kr
Log in for availability
1 * 750 Items
Product Details & Documents

Acetal polymer. Sleeved cassettes especially made to be used with Thermo Fisher printers. Cassettes stacked with tape should be used with Leica and Sakura inkjet printers.

  • The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage
  • Keep specimens safely submerged
  • Totally resistant to the chemical action of solvents used in histology laboratories
Specifications
  • Colour:
    Purple