Order Entry
Denmark
ContactUsLinkComponent
Box pp 60 lt folding blue,foldbox - polypropylene (pp) 60 l forstorage and transportation purposes temperature range from -30°c to + 80°c inner 570x370x310mm outer 600x400x310mm,blue colour. 1 * 1 items
Catalog # SEMA2802
Supplier:  SEMADENI
Box pp 60 lt folding blue,foldbox - polypropylene (pp) 60 l forstorage and transportation purposes temperature range from -30°c to + 80°c inner 570x370x310mm outer 600x400x310mm,blue colour. 1 * 1 items
Catalog # SEMA2802
Supplier:  SEMADENI
Frequently Bought Together