Burette 10Ml B Clear Grad=0.02Ml Str PTFE-Key Din En Iso 385, 1PC 1 * 2 items
HIRS3100360
Burette 10Ml B Clear Grad=0.02Ml Str PTFE-Key Din En Iso 385, 1PC 1 * 2 items
HIRS3100360
Frequently Bought Together
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection