Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cork stoppers

Cork stoppers

22 mm, 13 mm, 16 mm
Catalog # HECH41594016
Cork stoppers
Catalog # HECH41594016
Cork stoppers
Cork stoppers

22 mm, 13 mm, 16 mm

Price
1.880,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents

Cork stoppers designed for medical applications.

Specifications
  • Ø bottom:
    13 mm
  • Ø top:
    16 mm
  • Height:
    22 mm