Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tubing assemblies, PTFE

Tubing assemblies, PTFE

0,35 mm, 60 mm, White
Catalog # PERCB0501594
Supplier:   PerkinElmer
Tubing assemblies, PTFE
Catalog # PERCB0501594
Supplier:  PerkinElmer
Tubing assemblies, PTFE

0,35 mm, 60 mm, White

Price
1.130,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Made of PTFE.

  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Tubing int.Ø:
    0,35 mm
  • Colour:
    White
  • Length:
    60 mm
  • Seq:
    5
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.