Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tubing assemblies, PTFE

Tubing assemblies, PTFE

1,75 mm, 250 mm, Black
Catalog # PERCB0198099
Supplier:   PerkinElmer
Tubing assemblies, PTFE
Catalog # PERCB0198099
Supplier:  PerkinElmer
Tubing assemblies, PTFE

1,75 mm, 250 mm, Black

Price
1.940,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Made of PTFE.

  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Tubing int.Ø:
    1,75 mm
  • Colour:
    Black
  • Length:
    250 mm
  • Seq:
    6
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.