Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tubing assemblies, PTFE

Tubing assemblies, PTFE

1,0 mm, 700 mm, Blue
Catalog # PERCB0191059
Supplier:   PerkinElmer
Tubing assemblies, PTFE
Catalog # PERCB0191059
Supplier:  PerkinElmer
Tubing assemblies, PTFE

1,0 mm, 700 mm, Blue

Price
604,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Made of PTFE.

  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Tubing int.Ø:
    1,0 mm
  • Colour:
    Blue
  • Length:
    700 mm
  • Seq:
    3