Single channel pipettes, mechanical, fixed / variable volume, labopette®
100 - 1000 µl, 1 µl, ±0,8 %, <0,3 %, Blue, 10 - 1000 µl
Katalog # HIRS9475409
Leverandør: HIRSCHMANN
Single channel pipettes, mechanical, fixed / variable volume, labopette®
Katalog # HIRS9475409
Leverandør: HIRSCHMANN
Gå til:
Product Details & Documents
The reliable labopette® pipette features superior ergonomic characteristics, right down to the smallest detail. The entire working process is optimised to ensure non fatiguing work that places a minimum of stress on the joints.
- Volume configuration with digital locking feature
- With tip ejection and calibration option
- Tip seal and safety filter to prevent contamination
- Trigger grip ensures a firm but relaxed hold during pipetting
- Ergonomic design of the handle offers an equally firm, comfortable hold for large and small hands, right and left-handed users and hands with or without gloves
- Effective thermal insulation of the unit interior prevents hand heat penetrating into the interior of the unit
- Completely autoclavable without dismantling
Delivery information: Supplied with calibration tool, pipette holder, individual serial number and calibration certificate.
Specifikationer
- Volume:100 - 1000 µl
- Seq:07
- Increment:1 µl
- Accuracy:±0,8 %
- Imprecision:<0,3 %
- Recommended tips:Blue, 10 - 1000 µl
Accessories
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- HIRS9470010Safe-Cone filters, for labopette® variable 1000 - 5000 µl; fix 2000/5000 µlLog ind for tilgængelighed1 * 50 Items - 462,00 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- HIRS9470013Safe-Cone filters, for labopette® variable 2 - 20/5 - 50 µl; fix 20/25/50 µlLog ind for tilgængelighed1 * 50 Items - 462,00 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- HIRS9470011Safe-Cone filters, for labopette® elctronic 10 - 500/50 - 1000/500 - 5000/50 - 1200 µl; variable 100 - 1000 µl; fix 250/500/1000 µlLog ind for tilgængelighed1 * 50 Items - 462,00 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- HIRS9470012Safe-Cone filters, for labopette® variable 10 - 100/20 - 200 µl; fix 100/200 µlLog ind for tilgængelighed1 * 50 Items - 462,00 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- HIRS9980116DKD calibration certificate for labopette® fix 0,5 μl - 5 mlLog ind for tilgængelighed1 * 1 Items - 680,00 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- HIRS9980118DKD calibration certificate for labopette® 12-channel, 5 - 250 μlLog ind for tilgængelighed1 * 1 Items - 2.950,00 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- HIRS9470004Carousel stand for six mechanical pipettesLog ind for tilgængelighed1 * 1 Items - 1.380,00 kr
Produktinformation
- Katalog #VolumeTip colourFor use withTilgængelighedPrice
- HIRS94901010,2 - 10 µlTransparentHirschmann Laborgeräte, Gilson® pipettesLog ind for tilgængelighed1 * 1.000 Items - 267,00 kr
Produktinformation
- Katalog #VolumeTip colourFor use withTilgængelighedPrice
- HIRS94901021 - 200 µlYellowHirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettesLog ind for tilgængelighed1 * 1.000 Items - 178,00 kr
Produktinformation
- Katalog #VolumeTip colourFor use withTilgængelighedPrice
- HIRS9490103100 - 1000 µlBlueHirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettesLog ind for tilgængelighed1 * 1.000 Items - 198,00 kr
Produktinformation
- Katalog #VolumeTip colourFor use withTilgængelighedPrice
- HIRS94901061 - 250 µlTransparentlabopette® multichannel pipettesLog ind for tilgængelighed1 * 1.000 Items - 289,00 kr
Produktinformation
- Katalog #VolumeTip colourFor use withTilgængelighedPrice
- HIRS9490124100 - 5000 µlTransparentHirschmann Laborgeräte pipettesLog ind for tilgængelighed1 * 300 Items - 423,00 kr
Produktinformation
- Katalog #VolumeTip colourFor use withTilgængelighedPrice
- HIRS94902010,2 - 10 µlTransparentHirschmann Laborgeräte, Gilson® pipettesLog ind for tilgængelighed1 * 960 Items - 635,00 kr
Produktinformation
- Katalog #VolumeTip colourFor use withTilgængelighedPrice
- HIRS94902021 - 200 µlYellowHirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettesLog ind for tilgængelighed1 * 960 Items - 479,00 kr
Produktinformation
- Katalog #VolumeTip colourFor use withTilgængelighedPrice
- HIRS949020550 - 1200 µlTransparentHirschmann Laborgeräte pipettesLog ind for tilgængelighed1 * 960 Items - 1.190,00 kr
Produktinformation
- Katalog #VolumeTip colourFor use withTilgængelighedPrice
- HIRS94902061 - 250 µlTransparentlabopette® multichannel pipettesLog ind for tilgængelighed1 * 960 Items - 476,00 kr
Produktinformation
- Katalog #VolumeTip colourFor use withTilgængelighedPrice
- HIRS94901041 - 10000 µlTransparentHirschmann Laborgeräte pipettesLog ind for tilgængelighed1 * 100 Items - 423,00 kr
Produktinformation
- Katalog #Tip styleFor use withTilgængelighedPrice
- HIRS9490109Multibox, autoclavablerefill trays of pipette tips up to 300 µlLog ind for tilgængelighed1 * 1 Items - 227,00 kr
Produktinformation
- Katalog #Tip styleFor use withTilgængelighedPrice
- HIRS9490110Multibox, autoclavablerefill trays of pipette tips up to 1000 µlLog ind for tilgængelighed1 * 1 Items - 240,00 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- HIRS9470014Safe-Cone filters, for labopette® fix 10/20/25/250 µl; multi-channel 5,0 - 50/50 - 300 µl; electronic multi-channel 25 - 250/5 - 100 µlLog ind for tilgængelighed1 * 50 Items - 462,00 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- HIRS9980115DKD calibration certificate for labopette® variable volumes 0,5 μl - 5 ml / labopettorLog ind for tilgængelighed1 * 1 Items - 1.100,00 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- HIRS9980117DKD calibration certificate for labopette® 8-channel, 0,5 - 250 μlLog ind for tilgængelighed1 * 1 Items - 2.630,00 kr
Anbefalinger