VWR® Standard Micro Syringes
Standard micro syringe, 100 µl, 50 mm, Bevelled
Supplier: VWR Collection
Jump to:
Product Details & Documents
Standard micro syringes for GC with fixed needle.
- All parts in contact with liquid are made of stainless steel and neutral borosilicate glass
Length in ordering table refers to needle length.
Specifications
- Type of tip:Bevelled
- Volume:100 µl
- Description:Standard micro syringe
- Length:50 mm