Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR® Standard Micro Syringes
VWR® Standard Micro Syringes
Standard micro syringe, 10 µl, 50 mm, Bevelled
Catalog # 549-0199
Supplier:   VWR Collection
VWR® Standard Micro Syringes
Catalog # 549-0199
Supplier:  VWR Collection
VWR® Standard Micro Syringes
VWR® Standard Micro Syringes
Standard micro syringe, 10 µl, 50 mm, Bevelled
Price
324,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Standard micro syringes for GC with fixed needle.
  • All parts in contact with liquid are made of stainless steel and neutral borosilicate glass
Length in ordering table refers to needle length.
Specifications
  • Type of tip:
    Bevelled
  • Volume:
    10 µl
  • Description:
    Standard micro syringe
  • Length:
    50 mm