Order Entry
Denmark
ContactUsLinkComponent
Full option autoclave, fully automatic, microprocessor-controlled vertical laboratory autoclave with Cooling Sytem, Vacuum system,Printer and Swiss-plug, Chamberdimension (xL) 500 x 750 mm Volume: 160 liters 1 * 1 items
Catalog # TUTT5075ELV-FO-CH
Supplier:  TUTTNAUER
Full option autoclave, fully automatic, microprocessor-controlled vertical laboratory autoclave with Cooling Sytem, Vacuum system,Printer and Swiss-plug, Chamberdimension (xL) 500 x 750 mm Volume: 160 liters 1 * 1 items
Catalog # TUTT5075ELV-FO-CH
Supplier:  TUTTNAUER
Frequently Bought Together