0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Nicosulfuron certified 1 * 100 mg
EHERCA15515000
:
EHRENSTORFER
Nicosulfuron certified 1 * 100 mg
EHERCA15515000
:
EHRENSTORFER
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
Bottles, narrow neck, round, LDPE, with screw cap, 301 series
: Kautex
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
: VWR Chemicals
pH electrodes, SenTix® PLUS
: WTW
VWR®, Conductivity standard solutions
: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck