Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Turquoise
Catalog # 720-1927
Supplier: Simport Scientific
Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Catalog # 720-1927
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Acetal polymer. Sleeved cassettes especially made to be used with Thermo Fisher printers. Cassettes stacked with tape should be used with Leica and Sakura inkjet printers.
- The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage
- Keep specimens safely submerged
- Totally resistant to the chemical action of solvents used in histology laboratories
Specifications
- Colour:Turquoise
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



