Order Entry
Denmark
Orders LinkContactUsLinkComponent
Drum faucets
Drum faucets
Internal flame arrester, self-closing, wetted PTFE O-rings, 3/4"
Katalog # 119-0236
Leverandør:   JUSTRITE MANUFACTURING
Drum faucets
Katalog # 119-0236
Leverandør:  JUSTRITE MANUFACTURING
Drum faucets
Drum faucets
Internal flame arrester, self-closing, wetted PTFE O-rings, 3/4"
Price
550,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These faucets can be simply placed on your drum for leakproof dispensing of flammable liquids. The brass construction with a polytetrafluoroethylene (PTFE) seal ensures excellent chemical resistance.
  • Safe and easy dispensing
  • FM approved
The faucets feature an internal flame arrester that prevents flashback of fire into the drum for added safety.
Specifikationer
  • Thread:
    3/4"
  • Description:
    Internal flame arrester, self-closing, wetted PTFE O-rings
  • Seq:
    01
Anbefalinger