Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
VistaRak™, 250 µl, -, 5 racks, 192 each
Catalog # VIST4060-2004
Pipette tips
Catalog # VIST4060-2004
Pipette tips
Pipette tips
VistaRak™, 250 µl, -, 5 racks, 192 each
Price
564,00 kr
Log in for availability
1 * 960 Items
Product Details & Documents
These tips to maximise the exceptional performance and benefits of ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Specifications
  • Packed:
    5 racks, 192 each
  • Sterile:
    -
  • Volume:
    250 µl
  • Seq:
    05
  • Tip style:
    VistaRak™