Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stirrer guides

Stirrer guides

Stirrer guide, with cooling jacket and PTFE-insert, 29/32, 10,1 mm
Catalog # WITG1.475.029
Supplier:   witeg Labortechnik
Stirrer guides
Catalog # WITG1.475.029
Supplier:  witeg Labortechnik
Stirrer guides
Stirrer guides

Stirrer guide, with cooling jacket and PTFE-insert, 29/32, 10,1 mm

Price
2.390,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Stirrer guides are of different types.

With fused-on stirrer bearing.

GTS-vacuum-stirrer, with PTFE-insert and interchangeable PTFE-vacuum-seal, screw cap GL 25.

With PP-insert, 8 mm Ø and screw cap GL 25, made of melamine.

With cooling jacket and PTFE-insert, with interchangeable PTFE-vacuum seal and screw cap GL 25, made of melamine.

Specifications
  • NS:
    29/32
  • Description:
    Stirrer guide, with cooling jacket and PTFE-insert
  • Seq:
    05
  • Shaft Ø:
    10,1 mm