Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

pH buffer solutions, referenced at 20 °C, Fluka™

13,00, ±0.05, Glycine, 20 °C, Plastic bottle, 6×1 L, 6 L
Catalog # 33552-6X1L
Supplier:   Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33552-6X1L
CAS Number:  
pH buffer solutions, referenced at 20 °C, Fluka™

13,00, ±0.05, Glycine, 20 °C, Plastic bottle, 6×1 L, 6 L

CAS Number:  
Price
2.560,00 kr
Log in for availability
1 * 6 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Glycine
  • Pk:
    6 L
  • Reference temperature:
    20 °C
  • pH accuracy:
    ±0.05
  • Pk Info:
    6×1 L
  • pH value:
    13,00

Specifications