Order Entry
Denmark
Orders LinkContactUsLinkComponent
pH buffer solutions, referenced at 20 °C, Fluka™

pH buffer solutions, referenced at 20 °C, Fluka™

12,00, ±0.05, Phosphate, 20 °C, Plastic bottle, 6×1 L, 6 L
Catalog # 33651-6X1L
Supplier:   Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33651-6X1L
CAS Number:  
pH buffer solutions, referenced at 20 °C, Fluka™

12,00, ±0.05, Phosphate, 20 °C, Plastic bottle, 6×1 L, 6 L

CAS Number:  
Price
2.710,00 kr
Log in for availability
1 * 6 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Phosphate
  • Pk:
    6 L
  • Reference temperature:
    20 °C
  • pH accuracy:
    ±0.05
  • Pk Info:
    6×1 L
  • pH value:
    12,00

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.