Order Entry
Denmark
Orders LinkContactUsLinkComponent
pH buffer solutions, referenced at 20 °C, Fluka™

pH buffer solutions, referenced at 20 °C, Fluka™

12,00, ±0.05, Phosphate, 20 °C, Plastic bottle, 6×100 ml, 600 mL
Catalog # 33651-6X100ML
Supplier:   Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33651-6X100ML
CAS Number:  
pH buffer solutions, referenced at 20 °C, Fluka™

12,00, ±0.05, Phosphate, 20 °C, Plastic bottle, 6×100 ml, 600 mL

CAS Number:  
Price
1.310,00 kr
Log in for availability
1 * 600 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Phosphate
  • Pk:
    600 mL
  • Reference temperature:
    20 °C
  • pH accuracy:
    ±0.05
  • Pk Info:
    6×100 ml
  • pH value:
    12,00

Specifications