pH buffer solutions, referenced at 20 °C, Fluka™
11,00, ±0.05, Borate, 20 °C, Plastic bottle, 6×500 ml, 3.000 mL
Supplier: Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33650-6X500ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Borate
- Pk:3.000 mL
- Reference temperature:20 °C
- pH accuracy:±0.05
- Pk Info:6×500 ml
- pH value:11,00
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



