Chemiluminescent EMSA kit, LightShift™
LightShift™ EMSA optimisation and control kit (component of LightShift™ Chemiluminescent EMSA kit), 1 KIT
Catalog # PIER20148X
Supplier: Thermo Fisher Scientific
Chemiluminescent EMSA kit, LightShift™
Catalog # PIER20148X
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
LightShift™ Chemiluminescent EMSA Kit is a robust and sensitive system for performing electrophoretic mobility shift assays (EMSA) to identify and characterise protein-DNA binding interactions.
- Excellent for detecting low-abundance proteins in nuclear extracts
- Sensitivity that surpasses radioactive and digoxigenin methods
- Compatible with previously established binding conditions for popular DNA-protein interactions
- Includes EBNA Optimisation and Control Kit to help new users develop a working assay and understand the methods used to confirm binding interaction specificity
Delivery information: Kit includes LightShift™ EMSA Optimisation and Control Kit (Cat. No. 20148X) and Chemiluminescent Nucleic Acid Detection Module (Cat. No. 89880).
Specifications
Specifications
- Pk:1 KIT
- Description:LightShift™ EMSA optimisation and control kit (component of LightShift™ Chemiluminescent EMSA kit)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



