Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium hydroxide, BAKER, J.T.Baker®

Sodium hydroxide, BAKER, J.T.Baker®

25 kg
Catalog # 0405.9025
Corrosive
Supplier:   Avantor
Sodium hydroxide, BAKER, J.T.Baker®
Catalog # 0405.9025
Supplier:  Avantor
CAS Number:  1310-73-2
Sodium hydroxide, BAKER, J.T.Baker®

25 kg

CAS Number:  1310-73-2
Price
2.910,00 kr
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications

Specification Test Results

  • Assay:
    97.0-100.5 %
  • Appearance of solution:
    passes test
  • Carbonate (as Na2CO3):
    max. 2,0 %
  • Chlorides (as Cl):
    max. 50 ppm
  • Heavy Metals (as Pb):
    max. 20 ppm
  • Identification:
    passes test
  • Iron (Fe):
    max. 10 ppm
  • Sulfates (as SO4):
    max. 50 ppm
  • Additional Specification(s)::
    -
  • Potassium (K):
    passes test

Specifications

  • Pk:
    25 kg

Specifications